Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Lieferant: Simport Scientific
M525-2T M525-3T M525-4T M525-5T M525-6T M525-7T M525-8T M525-9T M525-10T M525-11T M525-12T
SIMPM525-2TEA 304 EUR
SIMPM525-2T SIMPM525-3T SIMPM525-4T SIMPM525-5T SIMPM525-6T SIMPM525-7T SIMPM525-8T SIMPM525-9T SIMPM525-10T SIMPM525-11T SIMPM525-12T
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Einbettkassetten Gewebeeinbettungskassetten
These cassettes are moulded from acetal and keep specimens safely submerged in liquid, and are resistant to the chemical action of most histological solvents.
  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica ink jet printers
  • Easy to remove disposable cover
Unisette™ histology cassette comes with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
The efficient flow-through slots maximise fluid exchange and ensureMore Product Information