Deep-Well-Platten, 96 quadratische Wells, Bioblock™
Lieferant: Simport Scientific
T110-10
SIMPT110-10EA 287 EUR
SIMPT110-10
Deep-Well-Platten, 96 quadratische Wells, Bioblock™
Mikrotiterplatten Deep-Well-Mikroplatten
PP. This 2,2 ml well capacity (2,1 ml when capped) plate is used mainly for compound storage and enzyme assays.
  • Conforms to SBS standards
  • Suitable to be used with Qiagen equipment
  • DMSO resistant
Lieferumfang: Packed in a bag of 4 plates.