Strips of 8 tubes
Lieferant: Simport Scientific
T110-15
SIMPT110-15EA 33.6 EUR
SIMPT110-15
Strips of 8 tubes
Röhrchen Mikroröhrchen
Made of PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Certified RNase, DNase, Pyrogen and DNA-free
These plates feature 600 µl wells for smaller volume applications. They include a deep well plate along with twelve eight-tube strips. The tube strips are removable and can also be ordered separately.