Deep well plates, 96-well, Bioblock™
Lieferant: Simport Scientific
T110-5
SIMPT110-5EA 266 EUR
SIMPT110-5
Deep well plates, 96-well, Bioblock™
Mikrotiterplatten Deep-Well-Mikroplatten
PP. The 1,2 ml capacity round bottom deep well plate (1 ml when capped) is easy to empty completely and ideal for culturing bacterial cells. It can withstand centrifugation up to 6000 ×g and is DMSO resistant.