Adapters for universal centrifuges
Lieferant: Simport Scientific
T330AD15 T330AD50
SIMPT330AD15EA 43.4 EUR
SIMPT330AD15 SIMPT330AD50
Adapters for universal centrifuges
Zentrifugen Tischzentrifugen
These universal centrifuge adapters are made of acetal plastic.
  • Reusable and freezable (–90 °C)