Human recombinant CCL1 (from HEK293 cells)
Lieferant: Sino Biological
10057-H01H-20
SINO10057-H01H-20EA 0 EUR
SINO10057-H01H-20
Human recombinant CCL1 (from HEK293 cells)
Proteine und Peptide
A DNA sequence encoding the mature form of human CCL1 (NP_002972.1) (Lys 24-Lys 96) was fused with the Fc region of human IgG1 at the N-terminus.