Human pro form of IL18 / Interleukin 18 / IGIF protein (GST Tag)(Inactive)
Lieferant: Sino Biological
10119-H09E-50
10119-H09E-20
SINO10119-H09E-50EA
362
EUR
SINO10119-H09E-50
SINO10119-H09E-20
Human pro form of IL18 / Interleukin 18 / IGIF protein (GST Tag)(Inactive)
Proteine und Peptide
A DNA sequence encoding the pro form of human IL18 (Q14116) (Met1-Asp193) was fused with the GST tag at the N-terminus.