Human IL2Ra / CD25 protein (ECD, Fc tag)
Lieferant: Sino Biological
10165-H02H-100
SINO10165-H02H-100EA 453 EUR
SINO10165-H02H-100
Human IL2Ra / CD25 protein (ECD, Fc tag)
Proteine und Peptide
A DNA sequence encoding the amino acid residues (Met 1-Cys 213) of the human IL2 receptor α chain (NP_000408.1) precursor was expressed with C-terminal fused human IgG1 Fc region.