Human HER3 / ErbB3 protein (ECD, his tag)
Lieferant: Sino Biological
10201-H08H-50
SINO10201-H08H-50EA
0
EUR
SINO10201-H08H-50
Human HER3 / ErbB3 protein (ECD, his tag)
Proteine und Peptide
A DNA sequence encoding the extracellular domain (Met 1-Thr 643) of human ErbB3 (NP_001973.2) precursor was expressed, with a C-terminal polyhistidine tag.