Human IL18BPa protein (His Tag)
Lieferant: Sino Biological
10357H08H-200
SINO10357H08H-200EA
0
EUR
SINO10357H08H-200
Human IL18BPa protein (His Tag)
Proteine und Peptide
A DNA sequence encoding the human IL18BPa (NP_001034748.1) (Met1-Gly194) was expressed with the polyhistidine tag at the C-terminus.