Human recombinant ERBB4 (from HEK293 cells), his tag
Lieferant: Sino Biological
10363-H08H-50
SINO10363-H08H-50EA 0 EUR
SINO10363-H08H-50
Human recombinant ERBB4 (from HEK293 cells), his tag
Proteine und Peptide
A DNA sequence encoding the human ERBB4 (NP_005226.1) (Met1-Arg649) with a C-terminal polyhistidine tag was expressed. Human and Rhesus ERBB4 sequences are identical.