Human IL6R / CD126 protein (his tag)
Lieferant: Sino Biological
10398-H08H-100
SINO10398-H08H-100EA 619 EUR
SINO10398-H08H-100
Human IL6R / CD126 protein (his tag)
Proteine und Peptide
A DNA sequence encoding the extracellular domain of human IL6R (NP_000556.1) corresponding to amino acid (Met1-Pro365) was expressed with a C-terminal polyhistidine tag.