Human TREM1 protein (his tag)
Lieferant: Sino Biological
10511-H08H-100
SINO10511-H08H-100EA 597 EUR
SINO10511-H08H-100
Human TREM1 protein (his tag)
Proteine und Peptide
A DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) was fused with the a polyhistidine tag at the C-terminus.