Human ULBP1 / N2DL1 protein (his tag)
Lieferant: Sino Biological
10679-H08H-100
SINO10679-H08H-100EA
520
EUR
SINO10679-H08H-100
Human ULBP1 / N2DL1 protein (his tag)
Proteine und Peptide
A DNA sequence encoding the mature form of human ULBP1 (NP_079494.1) (Met 1-Gly 216) was expressed, with a polyhistidine tag at the C-terminus.