Human CD80 / B7-1 protein (his tag)
Lieferant: Sino Biological
10698-H08H-100
SINO10698-H08H-100EA 344 EUR
SINO10698-H08H-100
Human CD80 / B7-1 protein (his tag)
Proteine und Peptide
A DNA sequence encoding the extracellular domain (Met 1-Asn 242) of human B7-1 (NP_005182.1) precursor was fused with a polyhistidine tag at the C-terminus.