Human recombinant DDR1 (from HEK293 cells), his tag
Lieferant: Sino Biological
10730-H08H-100
SINO10730-H08H-100EA
0
EUR
SINO10730-H08H-100
Human recombinant DDR1 (from HEK293 cells), his tag
Proteine und Peptide
A DNA sequence encoding the extracellular domain of human DDR1 isoform b (NP_001945.3) (Met 1-Thr 416) was expressed, with a C-terminal polyhistidine tag.