Human TIGIT / VSTM3 protein (Fc tag)
Lieferant: Sino Biological
10917-H02H-50
SINO10917-H02H-50EA
348
EUR
SINO10917-H02H-50
Human TIGIT / VSTM3 protein (Fc tag)
Proteine und Peptide
A DNA sequence encoding the human TIGIT (NP_776160.2) (Met1-Pro141) was expressed with the Fc region of human IgG1 at the C-terminus.