Human NOTCH1 protein (Fc tag)
Lieferant: Sino Biological
10954-H02H-50
SINO10954-H02H-50EA 0 EUR
SINO10954-H02H-50
Human NOTCH1 protein (Fc tag)
Proteine und Peptide
A DNA sequence encoding the human NOTCH1 (NP_060087.3) (Ala19-Gln526) was expressed with the Fc region of human IgG1 at the C-terminus.