SARS-CoV-2 (2019-nCoV) Nucleocapsid-His Recombinant Protein
Lieferant: Sino Biological
40588-V07E-100
SINO40588-V07E-100EA
808
EUR
SINO40588-V07E-100
SARS-CoV-2 (2019-nCoV) Nucleocapsid-His Recombinant Protein
Proteine und Peptide
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid Protein (YP_009724397.2) (Met1-Ala419(335Gly/Ala)) was expressed with a polyhistidine tag at the N-terminus.