Mouse TNFR1 / CD120a / TNFRSF1A Protein (Fc Tag)
Lieferant: Sino Biological
50496-M02H-100
SINO50496-M02H-100EA
0
EUR
SINO50496-M02H-100
Mouse TNFR1 / CD120a / TNFRSF1A Protein (Fc Tag)
Proteine und Peptide
A DNA sequence encoding the mouse TNFRSF1A (NP_ 035739.2) extracellular domain (Met 1-Ala 212) was fused with the Fc region of human IgG1 at the C-terminus.