Mouse IFNB1 / IFN-beta / Interferon beta Protein
Lieferant: Sino Biological
50708-MCCH-100
SINO50708-MCCH-100EA
0
EUR
SINO50708-MCCH-100
Mouse IFNB1 / IFN-beta / Interferon beta Protein
Proteine und Peptide
A DNA sequence encoding the mouse IFNB1 (P01575) (Met 1-Asn 182) was fused with five amino acids (DDDDK) at the C-terminus was expressed and purified.