Mouse HER2 / ErbB2 / CD340 Protein (His Tag)
Lieferant: Sino Biological
50714-M08H-100
SINO50714-M08H-100EA 0 EUR
SINO50714-M08H-100
Mouse HER2 / ErbB2 / CD340 Protein (His Tag)
Proteine und Peptide
A DNA sequence encoding the mouse ERBB2 (P70424) extracellular domain (Met 1-Thr 653) was expressed, with a C-terminal polyhistidine tag.