Mouse CHI3L1 / YKL40 / gp39 protein (His Tag)
Lieferant: Sino Biological
50929-M08H-50
SINO50929-M08H-50EA
0
EUR
SINO50929-M08H-50
Mouse CHI3L1 / YKL40 / gp39 protein (His Tag)
Proteine und Peptide
A DNA sequence encoding the extracellular domain of mouse CHI3L1 (Q61362) (Met 1-Ala 381) was expressed, with a C-terminal polyhistidine tag.