Mouse TNFSF14 / LIGHT / CD258 protein (His Tag)
Lieferant: Sino Biological
50980-M07B-100
SINO50980-M07B-100EA 503 EUR
SINO50980-M07B-100
Mouse TNFSF14 / LIGHT / CD258 protein (His Tag)
Proteine und Peptide
A DNA sequence encoding the mouse TNFSF14 (NP_062291.1) (Asp72-Val239) was expressed with a polyhistidine tag at the N-terminus.