Canine EGFR / HER1 / ErbB1 protein (His Tag)
Lieferant: Sino Biological
70026-D08B-50
SINO70026-D08B-50EA 0 EUR
SINO70026-D08B-50
Canine EGFR / HER1 / ErbB1 protein (His Tag)
Proteine und Peptide
A DNA sequence encoding the canine EGFR (XP_533073.3) (Met1-Ser618) was expressed with a C-terminal polyhistidine tag.