Rhesus IL-18 / IL-1F4 Protein (His Tag)
Lieferant: Sino Biological
90011-C07E-50
SINO90011-C07E-50EA
0
EUR
SINO90011-C07E-50
Rhesus IL-18 / IL-1F4 Protein (His Tag)
Proteine und Peptide
A DNA sequence encoding the rhesus IL-18 (NP_001028006.1) (Ala 2-Asp 193) was expressed with an N-terminal polyhistidine tag.