Sterilin™ Sample Bags, Homogeniser
Lieferant: Sterilin
S400
SLINS400EA 162 EUR
SLINS400
Sterilin™ Sample Bags, Homogeniser
Beutel Probenbeutel
Made of food-grade heavy-gauge polyethylene, these bags are double sealed for strength and reliability ensuring safe homogenisation of samples and are ideal for homogenising food samples prior to microbiological analysis. They are manufactured from food grade low density PE and are gamma irradiated to ensure the sterility.
  • Suitable for use in all leading homogeniser machines
  • Heavy gauge polyethylene for strength
  • Double-sealed for strength and reliability
  • Ensuring safe processing of samples
  • Manufactured from food-grade heavy-guage polyethylene for durability
  • Not available in all geographic regions