Disposable weighing boats, Sterilin®
Lieferant: Sterilin
WB30205 WB30314 WB30318 WB30324 WB30254 WB30334 WB30304 WB30317
611-3237EA 69.8 EUR
611-3237 611-3239 SLINWB30318 SLINWB30324 611-3238 611-3241 611-3622 SLINWB30317
Disposable weighing boats, Sterilin®
Schiffchen Wägeschiffchen
Polystyrene, white.
  • Radiused corners and sloping sides for complete sample transfer
  • Flat base for a secure firm rest on the balance pan
  • Suitable for coloured powders and samples
  • Diamond shape is particulrly flexible to allow the formation of a funnel for easy pouring to another vessel
Manufactured from high impact polystyrene with a hydrophobic surface that ensures no absorption of water from atmosphere or sample. The anti-staic version is made from PS material, modified to significantly reduce the charge relaxation time in comparison to standardMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable