Oligo(dT)₁₈ primer
Lieferant: Thermo Fisher Scientific
Oligo(dT)₁₈ primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Oligo(dT)₁₈ primer has been functionally tested in first strand cDNA synthesis.