Pipette stand, rotary, Twister™ universal 336
Lieferant: Socorex Isba
320.336W 320.336B 320.336Y 320.336G 320.336O 320.336Q 320.336R
SOCO320.336WEA 122.3 EUR
SOCO320.336W SOCO320.336B SOCO320.336Y SOCO320.336G SOCO320.336O SOCO320.336Q SOCO320.336R
Pipette stand, rotary, Twister™ universal 336
Pipettenständer
This rotary pipette work station fits six pipettes of most brands. It features a soft 360° rotation.
  • Ideal storage solution
  • Rotation enables quick instrument selection
  • Easy disassembling for simple maintenance
  • Available in seven translucent colours
  • Coloured discs are exchangeable