Pipette stand, 337
Lieferant: Socorex Isba
320.337B 320.337G 320.337M 320.337Y 320.337R
613-4830EA 99.4 EUR
613-4830 613-4831 613-0546 613-4832 SOCO320.337R
Pipette stand, 337
Pipettenständer
This space-saving pipette stand made from PA holds up to seven single channel pipettes of most brands.
  • Made of sturdy polyamid material, easy to clean
  • Very stable with anti-slip pads
  • Available in five pastel colours