Pipette charging stands
Lieferant: Socorex Isba
320.903.48 320.913.48
SOCO320.903.48EA 203 EUR
SOCO320.903.48 SOCO320.913.48
Pipette charging stands
Pipettenständer
These stands either allow the charging of up to three pipettes and/or battery packs or of three battery packs only.
  • Each position is fitted with a red/green charging LED
  • Rotary stands: Space saving