Cannula with spray nozzle, Dosys™
Lieferant: Socorex Isba
370.30500
SOCO370.30500EA 69.5 EUR
SOCO370.30500
Cannula with spray nozzle, Dosys™
Kanülen
This spray nozzle cannula fits on Luer-Lock self-refilling syringes.
Suitable where spraying and nebulising liquid is required.