Cleanroom Wipes, MicroPure
Lieferant: SONTARA
1000001885_U 1000003957_U 1000003838_U 1000001898 1000003980 1000001912
SONT1000001885_UEA 880 EUR
SONT1000001885_U SONT1000003957_U SONT1000003838_U SONT1000001898 SONT1000003980 SONT1000001912
Cleanroom Wipes, MicroPure
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
Sontara® Cleanroom wipes are specially designed for the challenging task of cleaning a cleanroom - ideal for electronic manufacturing, pharmaceutical, biotechnology applications and surface mount technology. These micro-level cleaning fabrics are 'virtually' lint-free and naturally absorbent, without binders or other additives.
  • Exceptionally clean wipe family with extremely low particle generation, low extractables and ion levels
  • Made with no binders, chemicals or adhesives
  • Special pattern for excellent pick-up
  • Excellent wet and dry strength
  • Solvent resistant
  • Extremely soft hand feel
  • Product traceability
The Sontara® cleanroom products help minimise the risk of contamination that meet all required levels of cleanliness and wiping needs.