Gel loading tips
Lieferant: Sorenson Bioscience
13810
SORE13810EA 330 EUR
SORE13810
Gel loading tips
Pipettenspitzen
These gel loading tips are available in different tip styles and packaging options.
  • Universal fit
  • Certified free of RNase/DNase, Human DNA, PCR inhibitors and pyrogens
  • Ideal for access to the bottom of narrow vessels and gel wells
  • MµltiFlex tips with 5 µl graduation, MiniFlex tips with 2 µl graduation
  • Outside orifice diameter of 0,37 mm for loading 0,4 mm thick gels
  • Outside orifice diameter of 0,17 mm for loading 0,2 mm thick gels
Standards: ISO 9001:2008