Barrier tips, MultiGuard3
Lieferant: Sorenson Bioscience
17370
SORE17370EA 124 EUR
SORE17370
Barrier tips, MultiGuard3
Pipettenspitzen
These natural coloured 200 µl MicroGuard3 PP barrier tips have a long sleek design and are available with low-binding surface technology.
  • Soft-fit top for easy eject and reliable multi-channel use
  • Graduations at 20, 50, 100, and 200 µl
  • Ultra-clear resin
  • Extended length: 77,8 mm
The optimal barrier placement and collar design allow the MultiGuard3 to be used with 20, 100 and 200 µl pipettors.