Standard Screw Caps for Microtubes
Lieferant: Sorenson Bioscience
390751CL
SORE390751CLEA 67.8 EUR
SORE390751CL
Standard Screw Caps for Microtubes
Verschlüsse Verschlüsse für Reagenz- und Probenröhrchen
Made of virgin polyethylene; medical grade.
  • Free of DNase, RNase and endotoxin
  • With O-ring and without seal
  • Non sterile
CE IVD marking.