Centrifuge, floor-standing, Sorvall™ RC4
Lieferant: SORVALL
75004473
SORV75004473EA 23994 EUR
SORV75004473
Centrifuge, floor-standing, Sorvall™ RC4
Zentrifugen Standzentrifugen
The RC4 is a flexible refrigerated centrifuge for blood banking and clinical applications.
  • SMARTspin™ Imbalance detection system automatically brakes unit if an excessive imbalance is detected, for example, due to tube leakage
  • Intuitive EASYcontrol™ II Soft-Touch keypad, Soft-Touch lid lock allows effortless opening and closing
  • Reduced maintenance with brushless induction drive
  • Very quiet operation at a noise level under 58 dBA
The versatile centrifuge offers optimal capacity for small to medium size blood processing centers with the ability to spin 4 or 8 blood bags in a wide range of bag configurations. A wide range of optional rotors makes it suitable for biochemistry, biotechnology, microbiology, pharmaceutical and blood processing labs.
Bestellinformation: Supplied without rotors which need to be ordered separately.