Nitroblau-Tetrazoliumchlorid (NBT)
Lieferant: SOUTHERN BIOTECHNOLOGY
|
Achtung
|
Synonyme:
5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylen)ditetrazoliumdichlorid, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylen)di(2-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid), 3,3'-(3,3'-Dimethoxy-4,4'-biphenylen)bis(2-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid)
0204-01
SOUT0204-01EA
0
EUR
SOUT0204-01
Nitroblau-Tetrazoliumchlorid (NBT)
Nitroblau-Tetrazoliumchlorid (NBT)
Formel:
C₄₀H₃₀Cl₂N₁₀O₆ Molecular Weight: 817,65 g/mol Schmelzpunkt: 200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00012159 CAS-Nummer: 298-83-9 EINECS: 206-067-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...