5-Brom-4-chlor-3-indolylphosphat p-Toluidinsalz (BCIP (p-Toluidinsalz), X-Phosphat)
Lieferant: SOUTHERN BIOTECHNOLOGY
|
Achtung
|
0205-01
SOUT0205-01EA
0
EUR
SOUT0205-01
5-Brom-4-chlor-3-indolylphosphat p-Toluidinsalz (BCIP (p-Toluidinsalz), X-Phosphat)
5-Brom-4-chlor-3-indolylphosphat p-Toluidinsalz (BCIP (p-Toluidinsalz), X-Phosphat)
Formel:
C₁₅H₁₅BrClN₂O₄P Lagertemperatur: Gefrierschrank |
MDL:
MFCD00040636 CAS-Nummer: 6578-06-9 EINECS: 229-506-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...