Examination light, Sovella® ProLite 28
Lieferant: TRESTON
861928-UK
SOVE861928-UKEA 0 EUR
SOVE861928-UK
Examination light, Sovella® ProLite 28
Beleuchtung Untersuchungsleuchten
This ergonomic lighting solution is environmentally friendly. Sovella® ProLite 28 is a standard light for workplaces where good general lighting is required but high accuracy is not necessary, for example packing stations.
  • Low energy consumption
  • All materials are recyclable
  • Prism diffuser shield which illuminates the work surface evenly and protects the fluorescent tube and technical components from dust
W×D×H: 1230×155×50 mm
Weight: 4,65 kg