ESD Saddle Chair
Lieferant: TRESTON
862010-00
SOVE862010-00EA 0 EUR
SOVE862010-00
ESD Saddle Chair
Stühle
Chairs with freedom of movement with ergonomic sitting position.
  • Adjustment for seat height and seat tilt
  • Abrasion resistance >50000
  • Martindale with ESD textile