LCD Screen Holder ESD with Double Arm
Lieferant: TRESTON
93049002P
SOVE93049002PEA 227 EUR
SOVE93049002P
LCD Screen Holder ESD with Double Arm
Laptopständer
LCD screen holder with a double arm, suitable for VESA 75 and 100 standards.
  • Epoxy powder-coated steel frame
  • Arm length 200+200 mm, thickness of the arm 60 mm