QuatreX Four-Leg Electric Bench Frame
Lieferant: TRESTON
QX18375-41
SOVEQX18375-41EA 1620 EUR
SOVEQX18375-41
QuatreX Four-Leg Electric Bench Frame
Möbel Tische und Werkbänke
Treston QuatreX electrical workbench is designed to offer supreme stability of a traditional four-legged workbench with the added advantage of extremely fast height adjustment.
  • Light grey
  • Motor adjustable
Treston QuatreX workbench gives the option to select either steel or aluminum uprights.
Total max loading capacity for the bench is 300 kg, for upright profiles total max loading capacity is 150 kg.
Lieferumfang: Supplied with 10 years warranty.