Acetic acid glacial_AcOH FCC
Lieferant: Spectrum Chemical
|
|
Synonyme:
Ethansäure
A1009-4LTPV
SPCMA1009-4LTPVEA
396
EUR
SPCMA1009-4LTPV
Acetic acid glacial_AcOH FCC
Essigsäure
Acetic Acid, Glacial, FCC is used in the pickling of vegetables and other foods. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Flame;Exclamation mark;Corrosion
UN2789
Formel:
H₃CCOOH Molecular Weight: 60.05 g/mol Siedepunkt: 118 Schmelzpunkt: 17 Dichte: 1,049 Flammpunkt: 38,5 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036152 CAS-Nummer: 64-19-7 UN: 2789 ADR: 8.3 II |
|
Spezifikation Testergebnisse
Assay | 99.5 - 100.5% |
Solidification Point | 15.6°C min. |
Nonvolatile Residue | max. 0.005 % |
Lead (Pb) | max. 0.5 mg/kg |
Readily Oxidizable Substances | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...