Aluminium potassium sulphate dodecahydrate 99,5-100,5%, Kristalle FCC

Lieferant: Spectrum Chemical

Synonyme: Alumen, Kalialaun, Aluminiumkaliumbis(sulfat) Dodecahydrat

A1107-12KG
SPCMA1107-12KGEA 1710 EUR
SPCMA1107-12KG
Aluminium potassium sulphate dodecahydrate 99,5-100,5%, Kristalle FCC
Aluminiumkaliumsulfat Dodecahydrat

Aluminum Potassium Sulfate, Crystal, FCC is commonly used as an additive to baking powder to provide a second leavening phase at high temperatures. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: AlK(SO₄)₂·12H₂O
Molecular Weight: 474,39 g/mol
Schmelzpunkt: 92,5 °C/198,5 °F
Dichte: 1,757
Lagertemperatur: Raumtemperatur
MDL: MFCD00149143
CAS-Nummer: 7784-24-9

Bestellen

Spezifikation Testergebnisse

Assay [Alk(SO₄)₂[-12H₂O] 99.5-100.5%
Ammonium Salts To Pass Test
Fluoride max. 0.003%
Lead (Pb) max. 3 mg/kg
Selenium (Se) max. 0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR