Ammoniumchlorid 99,0, granular FCC

Lieferant: Spectrum Chemical

Synonyme: Salmiak

A1167-2.5KG
SPCMA1167-2.5KGEA 554 EUR
SPCMA1167-2.5KG
Ammoniumchlorid 99,0, granular FCC
Ammoniumchlorid

Ammonium Chloride, Granular, FCC is an ingredient in nutritive media for yeasts and many microorganisms. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formel: NH₄Cl
Molecular Weight: 53.49 g/mol
Schmelzpunkt: 335
Lagertemperatur: Raumtemperatur
MDL: MFCD00143604
CAS-Nummer: 12125-02-9
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (Dried Basis) 99.0 % min.
Lead (Pb) max.4 mg/kg
Loss On Drying max. 0.5 %
Identification To Pass Test

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR