N-Acetyl-β-D-glucosamin

Lieferant: Spectrum Chemical

Synonyme: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

A1269-25KGBL
SPCMA1269-25KGBLEA 0 EUR
SPCMA1269-25KGBL
N-Acetyl-β-D-glucosamin
N-Acetyl-β-D-glucosamin
Formel: C₈H₁₅NO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00136044
CAS-Nummer: 7512-17-6
EINECS: 231-368-2

Bestellen

Spezifikation Testergebnisse

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR