L(+)-Asparagine monohydrate ≥98,0% FCC

Lieferant: Spectrum Chemical

Synonyme: (S)-(+)-Asparagin Monohydrat, (S)-(+)-2-Amino-1,4-butandisäure-4-amid Monohydrat, L-Asparagin Monohydrat, L(+)-2-Amino-1,4-butandisäure-4-amid Monohydrat, H-Asn-OH.H2O

A1373-100GM
SPCMA1373-100GMEA 168 EUR
SPCMA1373-100GM
L(+)-Asparagine monohydrate ≥98,0% FCC
L(+)-Asparagin Monohydrat

L-Asparagine, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₄H₈N₂O₃·1H₂O
Molecular Weight: 150,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151038
CAS-Nummer: 5794-13-8

Bestellen

Spezifikation Testergebnisse

Assay (Dried Basis) 98.0 - 101.5%
Specific Rotation [A]D +33.0 to +36.5
Loss On Drying 11.5 - 12.5 %
Lead (Pb) max. 5 mg/kg
Residue On Ignition max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR