Ammonium sulphide -Lösung ACS

Lieferant: Spectrum Chemical

A1404-100ML
SPCMA1404-100MLEA 158 EUR
SPCMA1404-100ML
Ammonium sulphide -Lösung ACS
Ammonium sulphide

Ammonium Sulfide Solution, Reagent, ACS is an unstable salt with the formula (NH4)2S. Common uses of AMMONIUM SULFIDE are industrial applications including photo developing and textile manufacturing.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formel: H₈N₂S
Molecular Weight: 68.14 g/mol
Schmelzpunkt: -18
Dichte: 0,997
Lagertemperatur: Raumtemperatur
MDL: MFCD00010892
CAS-Nummer: 12135-76-1
UN: 2683
ADR: 86.13 II
Merck Index: 13,00560

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR